Learn More
SB-399885 hydrochloride, MedChemExpress
Product Code. 16442866
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16442866
Brand: Medchem Express HY103099
MedChemExpress SB-399885 hydrochloride is a 5-HT6 receptor antagonist.
SB-399885 hydrochloride is a 5-HT6 receptor antagonist.
Specifications
Chemical Name or Material SB-399885 hydrochloride
CAS 402713-81-9
Color White
Purity Grade Notes Research
Molecular Formula C18H22Cl3N3O4S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (207.12 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.81 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC=C(OC)C(N2CCNCC2)=C1)(NC3=CC(Cl)=CC(Cl)=C3OC)=O.[H]Cl
Molecular Weight (g/mol) 482.81
Formula Weight 482.81
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation