Learn More
DDR1-IN-1, MedChemExpress
Product Code. 16432097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16432097
Brand: Medchem Express HY13979
MedChemExpress DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM; 4-fold less potent for DDR2 (IC50 = 413 nM).
DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM
Specifications
Chemical Name or Material DDR1-IN-1
CAS 1449685-96-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H31F3N4O3
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (180.97 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C)C(OC2=CC3=C(NC(C3)=O)C=C2)=C1)C4=CC=C(CN5CCN(CC)CC5)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 552.59
Formula Weight 552.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease