Learn More
Abemaciclib methanesulfonate, MedChemExpress
Product Code. 16431806
Encompass_Preferred
Click to view available options
Quantity:
5 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
5mg
Product Code. 16431806
Brand: Medchem Express HY16297
MedChemExpress Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Specifications
Chemical Name or Material Abemaciclib methanesulfonate
CAS 1231930-82-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H36F2N8O3S
Quantity 500 mg
Synonym LY2835219 methanesulfonate
Solubility Information H2O : 125 mg/mL (207.40 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (16.59 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O
Molecular Weight (g/mol) 602.7
Formula Weight 602.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease