Learn More
Levomefolic acid, MedChemExpress
Product Code. 16422307
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16422307
Brand: Medchem Express HY14781
MedChemExpress Levomefolic acid (5-MTHF) is an orally active, brain-penetrant natural active form of folic acid and is one of the most widely used folic acid food supplements.
Levomefolic acid (5-MTHF) is an orally active, brain-penetrant natural active form of folic acid and is one of the most widely used folic acid food supplements.
Specifications
Chemical Name or Material Levomefolic acid
Physical Form Solid
CAS 31690-09-2
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H25N7O6
For Use With (Application) Cancer-programmed cell death
Synonym 5-MTHF
Solubility Information H2O : 5 mg/mL (10.88 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.18 mM; Need ultrasonic)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(C1=CC=C(NC[C@@H]2N(C)C3=C(NC(N)=NC3=O)NC2)C=C1)=O
Molecular Weight (g/mol) 459.46
Formula Weight 459.46
Percent Purity 98.0%
Grade Research