Learn More
ETP-46321, MedChemExpress
Product Code. 16415836
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415836 100 mg 100mg
16477827 5 mg 5mg
16445108 10 mg 10mg
16411347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16415836 Supplier Medchem Express Supplier No. HY12340
MedChemExpress ETP-46321 is a potent and orally bioavailable PI3Kα and PI3Kδ inhibitor with Kiapps of 2.3 and 14.2 nM, respectively.
ETP-46321 is a potent and orally bioavailable PI3Kα and PI3Kδ inhibitor with Kiapps of 2.3 and 14.2 nM, respectively.
Specifications
Chemical Name or Material ETP-46321
CAS 1252594-99-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H27N9O3S
Quantity 100 mg
Solubility Information DMSO : ≥ 33 mg/mL (69.69 mM)
SMILES NC(N=C1)=NC=C1C2=CN3C=C(CN4CCN(S(=O)(C)=O)CC4)N=C3C(N5CCOCC5)=N2
Molecular Weight (g/mol) 473.55
Formula Weight 473.55
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease