Learn More
Vitamin D3, MedChemExpress
Product Code. 16411519
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Product Code. 16411519
Brand: Medchem Express HY15398
MedChemExpress Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occuring form of vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells.
Vitamin D3 (Cholecalciferol
Specifications
Chemical Name or Material Vitamin D3
CAS 67-97-0
Color White
Purity Grade Notes Research
Molecular Formula C27H44O
Quantity 1 g
Synonym Cholecalciferol Colecalciferol
Solubility Information DMSO : 100 mg/mL (259.98 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCCC(C)C)([H])/C(CCC1)=C/C=C3C(CC[C@H](O)C\3)=C
Molecular Weight (g/mol) 384.64
Formula Weight 384.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death