Learn More
AZ505 ditrifluoroacetate, MedChemExpress
Product Code. 16408967
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16408967
Brand: Medchem Express HY15226A
MedChemExpress AZ505 ditrifluoroacetate is a potent and selective SMYD2 inhibitor with IC50 of 0.12 μM.
AZ505 ditrifluoroacetate is a potent and selective SMYD2 inhibitor with IC50 of 0.12 μM.
Specifications
Chemical Name or Material AZ505 ditrifluoroacetate
CAS 1035227-44-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H40Cl2F6N4O8
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (155.17 mM)
SMILES O=C(N(C1CCCCC1)CCNCCC2=C(OCC(N3)=O)C3=C(O)C=C2)CCNCCC4=CC=C(Cl)C(Cl)=C4.O=C(O)C(F)(F)F.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 805.59
Formula Weight 805.59
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death