Learn More
(R)-Filanesib, MedChemExpress
Product Code. 16408947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408947 5 mg 5mg
16452467 50 mg 50mg
16416666 100 mg 100mg
16416358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16408947 Supplier Medchem Express Supplier No. HY15187A
MedChemExpress (R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM.
(R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM.
Specifications
Chemical Name or Material (R)-Filanesib
CAS 885060-08-2
Color White
Purity Grade Notes Research
Molecular Formula C20H22F2N4O2S
Quantity 5 mg
Synonym (R)-ARRY-520
Solubility Information DMSO : ≥ 100 mg/mL (237.82 mM)
SMILES O=C(N1[C@](C2=CC=CC=C2)(CCCN)SC(C3=CC(F)=CC=C3F)=N1)N(OC)C
Molecular Weight (g/mol) 420.48
Formula Weight 420.48
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death