Learn More
Canagliflozin hemihydrate, MedChemExpress
Product Code. 16408936
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408936 100 mg 100mg
16494707 50 mg 50mg
16461238 5 mg 5mg
16428808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16408936 Supplier Medchem Express Supplier No. HYI0383
MedChemExpress Canagliflozin hemihydrate (JNJ28431754 hemihydrate) is a selective SGLT2 inhibitor with IC50s of 2 nM, 3.7 nM, and 4.4 nM for mSGLT2, rSGLT2, and hSGLT2 in CHOK cells, respectively.
Canagliflozin hemihydrate (JNJ28431754 hemihydrate) is a selective SGLT2 inhibitor with IC50s of 2 nM, 3.7 nM, and 4.4 nM for mSGLT2, rSGLT2, and hSGLT2 in CHOK cells, respectively.
Specifications
Chemical Name or Material Canagliflozin hemihydrate
CAS 928672-86-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26FO5.5S
Quantity 100 mg
Synonym JNJ 28431754 hemihydrate
Solubility Information DMSO : ≥ 100 mg/mL (220.50 mM)
SMILES CC1=CC=C([C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)C=C1CC3=CC=C(C4=CC=C(F)C=C4)S3.[0.5H2O]
Molecular Weight (g/mol) 453.52
Formula Weight 453.52
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism