Learn More
I-BET151, MedChemExpress
Product Code. 16408277
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16408277
Brand: Medchem Express HY13235
MedChemExpress I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.
I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.
Specifications
Chemical Name or Material I-BET151
CAS 1300031-49-5
Color White
Purity Grade Notes Research
Molecular Formula C23H21N5O3
Quantity 5 mg
Synonym GSK1210151A
Solubility Information DMSO : ≥ 100 mg/mL (240.71 mM)
SMILES C[C@@H](N1C(C(C=C(OC)C(C2=C(C)ON=C2C)=C3)=C3N=C4)=C4NC1=O)C5=CC=CC=N5
Molecular Weight (g/mol) 415.44
Formula Weight 415.44
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death