Learn More
Letrozole, MedChemExpress
Product Code. 16406426
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16406426
Brand: Medchem Express HY14248
MedChemExpress Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.
Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.
Specifications
Chemical Name or Material Letrozole
Physical Form Powder
CAS 112809-51-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H11N5
For Use With (Application) Cancer-programmed cell death
Synonym CGS 20267
Solubility Information DMSO : 50 mg/mL (175.25 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C(N2C=NC=N2)C3=CC=C(C#N)C=C3)C=C1
Molecular Weight (g/mol) 285.3
Formula Weight 285.3
Percent Purity 99.79%
Grade Research