Learn More
Staurosporine, MedChemExpress
Product Code. 16406328
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
Product Code. 16406328
Brand: Medchem Express HY15141
MedChemExpress Staurosporine is a potent, ATP-competitive and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine also inhibits TAOK2 with an IC50 of 3 μM. Staurosporine is an apoptosis inducer.
Staurosporine is a potent, ATP-competitive and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine also inhibits TAOK2 with an IC50 of 3 μM. Staurosporine is an apoptosis inducer.
Specifications
Chemical Name or Material Staurosporine
CAS 62996-74-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H26N4O3
Quantity 10 mg
Synonym Antibiotic AM-2282 STS AM-2282
Solubility Information DMSO : 62.5 mg/mL (133.97 mM; Need ultrasonic)
SMILES O=C(NC1)C2=C1C3=C(C4=C2C5=C(C=CC=C5)N4[C@H]6C[C@@H](NC)[C@@H](OC)[C@]7(C)O6)N7C8=CC=CC=C83
Molecular Weight (g/mol) 466.53
Formula Weight 466.53
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease