Learn More
AMG319, MedChemExpress
Product Code. 16405448
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16405448
Brand: Medchem Express HY12948
MedChemExpress AMG319 is a potent and selective PI3Kδ kinase inhibitor with IC50 of 18 nM.
AMG319 is a potent and selective PI3Kδ kinase inhibitor with IC50 of 18 nM.
Specifications
Chemical Name or Material AMG319
CAS 1608125-21-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H16FN7
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (129.74 mM; Need ultrasonic)
SMILES C[C@H](NC1=C2N=CNC2=NC=N1)C3=CC4=CC=C(F)C=C4N=C3C5=NC=CC=C5
Molecular Weight (g/mol) 385.4
Formula Weight 385.4
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease