Learn More
Nobiletin, MedChemExpress
Product Code. 16404777
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16404777
Brand: Medchem Express HYN0155
MedChemExpress Nobiletin is a poly-methoxylated flavone from the citrus peel that improves memory loss. Nobiletin is a retinoid acid receptor-related orphan receptors (RORs) agonist. Nobiletin can reduce reactive oxygen species (ROS) levels in differentiated C2C12 myotubes and has anti-inflammation and anti-cancer properties, including anti-angiogenesis, anti-proliferation, anti-metastasis and induced apoptosis.
Nobiletin is a poly-methoxylated flavone from the citrus peel that improves memory loss. Nobiletin is a retinoid acid receptor-related orphan receptors (RORs) agonist. Nobiletin can reduce reactive oxygen species (ROS) levels in differentiated C2C12 myotubes and has anti-inflammation and anti-cancer properties, including anti-angiogenesis, anti-proliferation, anti-metastasis and induced apoptosis.
Specifications
Chemical Name or Material Nobiletin
CAS 478-01-3
Color White
Purity Grade Notes Research
Molecular Formula C21H22O8
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (124.26 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=C(OC)C(OC)=C(OC)C(OC)=C13
Molecular Weight (g/mol) 402.39
Formula Weight 402.39
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease