Learn More
GSK 690 Hydrochloride, MedChemExpress
Description
GSK 690 (Hydrochloride) is a reversible inhibitor of lysine specific demethylase 1 (LSD1), with a Kd value of 9 nM and a biochemical IC50 of 37 nM.
Specifications
Specifications
| Chemical Name or Material | GSK 690 Hydrochloride |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H24ClN3O |
| Quantity | 50 mg |
| Solubility Information | DMSO : 125 mg/mL (307.94 mM; Need ultrasonic) |
| SMILES | N#CC1=CC=C(C2=CC(OC[C@H]3CNCC3)=CN=C2C4=CC=C(C)C=C4)C=C1.Cl |
| Molecular Weight (g/mol) | 405.92 |
| Formula Weight | 405.92 |
| Percent Purity | 99.16% |
| Show More |