Learn More
Invitrogen™ HMGN2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA585030
Product Code. 16391975
  • £478.00 / 100µL
Description
Immunogen sequence: PKRKAKGDAK GDKAKVKDEP Q Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 90%.
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HMGN2
Polyclonal
Unconjugated
HMGN2
high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; high mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17
Rabbit
Antigen affinity chromatography
RUO
3151
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P05204
HMGN2
Recombinant protein corresponding to Human HMGN2.