Learn More
Invitrogen™ MRPL14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585010
This item is not returnable.
View return policy
Description
Immunogen sequence: FSTTGSLSAI QKMTRVRVVD NSALGNSPYH RAPRCIHVYK KNGVGKVGDQ ILLAIK Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 86%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
Specifications
| MRPL14 | |
| Polyclonal | |
| Unconjugated | |
| Mrpl14 | |
| 1110006I11Rik; 39S ribosomal protein L14, mitochondrial; 39S ribosomal protein L32, mitochondrial; AI846816; L14mt; L32mt; Mitochondrial large ribosomal subunit protein uL14m; mitochondrial ribosomal protein L14; mitochondrial ribosomal protein L32; Mrpl14; MRP-L14; MRPL32; MRP-L32; RMPL32; Rpml32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P1L8 | |
| Mrpl14 | |
| Recombinant protein corresponding to Human MRPL14. Recombinant protein control fragment (Product #RP-108098). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |