Learn More
Invitrogen™ IGF2BP3 Monoclonal Antibody (C2-E4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA532838
Product Code. 16386244
  • £439.00 / 100µL
Description
IGF2BP3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some malignancies. IMP-3 is expressed in malignant melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of malignant melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGF2BP3
Monoclonal
2 mg/mL
PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4
O00425, Q9CPN8
IGF2BP3
Recombinant protein within Human IMP-3 aa 352-579.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
C2-E4
Unconjugated
IGF2BP3
2610101N11Rik; AA522010; Ab2-255; AL022933; AU045931; cancer/testis antigen 98; CT98; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; insulin-like growth factor 2, binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; Koc13; mimp3; mIMP-3; Neilsen; RGD1306512; VICKZ family member 3; Vickz3
Mouse
Protein A
RUO
10643, 140488, 312320
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications