Learn More
Invitrogen™ ADAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583523
This item is not returnable.
View return policy
Description
Immunogen sequence: YLLHQLQLAA TLKEDSIFVP GTQKGVWKLR RDLIFVFFSS HTPCGDASII PMLEFEDQPC CPVFRNWAHN SSVEASSNLE APGNERKCED PD Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 75%.
This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA.
Specifications
| ADAT1 | |
| Polyclonal | |
| Unconjugated | |
| ADAT1 | |
| Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23536 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUB4 | |
| ADAT1 | |
| Recombinant protein corresponding to Human ADAT1. Recombinant protein control fragment (Product #RP-98539). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |