Learn More
Invitrogen™ ZNRD1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA585046
Product Code. 16351995
  • £478.00 / 100µL
Description
Immunogen sequence: GTAMPMSVEE GPECQGPVVD RRCPRCGHEG MAYHTRQMRS ADEGQTVFYT CTNCKFQEKE DS Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 87%.
This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNRD1
Polyclonal
Unconjugated
ZNRD1
1110014N07Rik; DNA-directed RNA polymerase I subunit RPA12; HTEX6; HTEX-6; hZR14; nuclear RNA polymerase I small specific subunit; nuclear RNA polymerase I small specific subunit Rpa12 like; RNA polymerase I small specific subunit Rpa12; Rpa12; Tctex6; tctex-6; TEX6; transcription-associated zinc ribbon protein; zinc ribbon domain containing 1; zinc ribbon domain containing, 1; zinc ribbon domain-containing protein 1; ZNRD1; ZR14
Rabbit
Antigen affinity chromatography
RUO
30834
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9P1U0
ZNRD1
Recombinant protein corresponding to Human ZNRD1. Recombinant protein control fragment (Product #RP-108697).