Learn More
Invitrogen™ HELIOS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585425
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Rhesus Monkey (100%). Positive Control: Jurkat, Jurkat NE. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
| HELIOS | |
| Polyclonal | |
| Unconjugated | |
| IKZF2 | |
| A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22779, 22807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P81183, Q9UKS7 | |
| IKZF2 | |
| Recombinant protein encompassing a sequence within the center region of human HELIOS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |