Learn More
Uricase, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native Uricase (Bacillus).
Brand: Abnova PAB28038.100ug
This item is not returnable.
View return policy
Specifications
| Uricase | |
| Polyclonal | |
| Goat polyclonal antibody raised against native Uricase (Bacillus). | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| ABC3735 | |
| Native purified Bacillus Uricase. | |
| RUO | |
| 3202700 | |
| Lyophilized |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user. | |
| ABC3735 | |
| Goat | |
| 100 μg | |
| Primary | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |