Learn More
Abnova™ Human OR2J2 Full-length ORF (NP_112167.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16188412
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16188412
Brand: Abnova™ H00026707P01.10ug
Used for AP, Array, ELISA, WB-Re
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]
Sequence: MMIKKNASSEDFFILLGFSNWPQLEVVLFVVILIFYLMTLTGNLFIIILSYVDSHLHTPMYFFLSNLSFLDLCHTTSSIPQLLVNLRGPEKTISYAGCMVQLYFVLALGIAECVLLVVMSYDRYVAVCRPLHYTVLMHPRFCHLLAAASWVIGFTISALHSSFTFWVPLCGHRLVDHFFCEVPALLRLSCVDTHANELTLMVMSSIFVLIPLILILTAYGAIARAVLSMQSTTGLQKVFRTCGAHLMVVSLFFIPVMCMYLQPPSENSPDQGKFIALFYTVVTPSLNPLIYTLRNKHVKGAAKRLLGWEWGK
Specifications
Accession Number NP_112167.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26707
Molecular Weight (g/mol) 61.6kDa
Name OR2J2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MMIKKNASSEDFFILLGFSNWPQLEVVLFVVILIFYLMTLTGNLFIIILSYVDSHLHTPMYFFLSNLSFLDLCHTTSSIPQLLVNLRGPEKTISYAGCMVQLYFVLALGIAECVLLVVMSYDRYVAVCRPLHYTVLMHPRFCHLLAAASWVIGFTISALHSSFTFWVPLCGHRLVDHFFCEVPALLRLSCVDTHANELTLMVMSSIFVLIPLILILTAYGAIARAVLSMQSTTGLQKVFRTCGAHLMVVSLFFIPVMCMYLQPPSENSPDQGKFIALFYTVVTPSLNPLIYTLRNKHVKGAAKRLLGWEWGK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC119134/MGC119135/MGC119136/MGC119137/OR6-19/OR6-8/OR6.3.8/ORL684/dJ80I19.4/hs6M1-6
Common Name OR2J2
Gene Symbol OR2J2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid