Learn More
SLC17A2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC17A2.
Brand:  Abnova PAB23256.100uL
Product Code. 16185810
  • £535.00 / 100µL
Description
Sequence: AIIAMVNTTQQQGLSNASTEGPVADAFNNSSISIKEFDTKASVYQWSPETQG
Specifications
Specifications
SLC17A2
Polyclonal
Rabbit polyclonal antibody raised against recombinant SLC17A2.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
MGC138238/NPT3
Rabbit
Antigen affinity purification
RUO
10246
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
SLC17A2
SLC17A2
Recombinant protein corresponding to amino acids of human SLC17A2.