Learn More
Abnova™ Human GAN Full-length ORF (NP_071324.1, 1 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16182292
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16182292
Brand: Abnova™ H00008139P01.25ug
Used for AP, Array, ELISA, WB-Re
This gene encodes a member of the cytoskeletal BTB/kelch (Broad-Complex, Tramtrack and Bric a brac) repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy (GAN). [provided by RefSeq]
Sequence: MAEGSAVSDPQHAARLLRALSSFREESRFCDAHLVLDGEEIPVQKNILAAASPYIRTKLNYNPPKDDGSTYKIELEGISVMVMREILDYIFSGQIRLNEDTIQDVVQAADLLLLTDLKTLCCEFLEGCIAAENCIGIRDFALHYCLHHVHYLATEYLETHFRDVSSTEEFLELSPQKLKEVISLEKLNVGNERYVFEAVIRWIAHDTEIRKVHMKDVMSALWVSGLDSSYLREQMLNEPLVREIVKECSNIPLSQPQQGEAMLANFKPRGYSECIVTVGGEERVSRKPTAAMRCMCPLYDPNRQLWIELAPLSMPRINHGVLSAEGFLFVFGGQDENKQTLSSGEKYDPDANTWTALPPMNEARHNFGIVEIDGMLYILGGEDGEKELISMECYDIYSKTWTKQPDLTMVRKIGCYAAMKKKIYAMGGGSYGKLFESVECYDPRTQQWTAICPLKERRFGAVACGVAMELYVFGGVRSREDAQGSEMVTCKSEFYHDEFKRWIYLNDQNLCIPASSSFVYGAVPIGASIYVIGDLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSP
Specifications
Accession Number NP_071324.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8139
Molecular Weight (g/mol) 94kDa
Name GAN (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAEGSAVSDPQHAARLLRALSSFREESRFCDAHLVLDGEEIPVQKNILAAASPYIRTKLNYNPPKDDGSTYKIELEGISVMVMREILDYIFSGQIRLNEDTIQDVVQAADLLLLTDLKTLCCEFLEGCIAAENCIGIRDFALHYCLHHVHYLATEYLETHFRDVSSTEEFLELSPQKLKEVISLEKLNVGNERYVFEAVIRWIAHDTEIRKVHMKDVMSALWVSGLDSSYLREQMLNEPLVREIVKECSNIPLSQPQQGEAMLANFKPRGYSECIVTVGGEERVSRKPTAAMRCMCPLYDPNRQLWIELAPLSMPRINHGVLSAEGFLFVFGGQDENKQTLSSGEKYDPDANTWTALPPMNEARHNFGIVEIDGMLYILGGEDGEKELISMECYDIYSKTWTKQPDLTMVRKIGCYAAMKKKIYAMGGGSYGKLFESVECYDPRTQQWTAICPLKERRFGAVACGVAMELYVFGGVRSREDAQGSEMVTCKSEFYHDEFKRWIYLNDQNLCIPASSSFVYGAVPIGASIYVIGDLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ38059/GAN1/KLHL16
Common Name GAN
Gene Symbol GAN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid