Learn More
REM2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand:  Abnova H00161253-B01P.50ug
Product Code. 16178903
  • £303.00 / 50µg
Description
Sequence: MDTETTALCPSGSRRASPPGTPTPEADATLLKKSEKLLAELDRSGLPSAPGAPRRRGSMPVPYKHQLRRAQAVDELDWPPQASSSGSSDSLGSGEAAPAQKDGIFKVMLVGESGVGKSTLAGTFGGLQGDSAHEPENPEDTYERRIMVDKEEVTLVVYDIWEQGDAGGWLRDHCLQTGDAFLIVFSVTDRRSFSKVPETLLRLRAGRPHHDLPVILVGNKSDLARSREVSLEEGRHLAGTLSCKHIETSAALHHNTRELFEGAVRQIRLRRGRNHAGGQRPDPGSPEGPAPPARRESLTKKAKRFLANLVPRNAKFFKQRSRSCHDLSVL
Specifications
Specifications
REM2
Polyclonal
In 1x PBS, pH 7.4
BC035663
REM2
FLJ38964 (AAH35663, 1 a.a. ∼ 330 a.a) full-length human protein.
RUO
161253
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Immunofluorescence, Western Blot
Unconjugated
RAS (RAD and GEM)-like GTP binding 2
FLJ38964