Learn More
GRIK3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00002899-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MTAPWRRLRSLVWEYWAGLLVCAFWIPDSRGMPHVIRIGGIFEYADGPNAQVMNAEEHAFRFSANIINRNRTLLPNTTLTYDIQRIHFHDSFEATKKACDQLALGVVAIFGPSQGSCTNAVQSICNALEVPHIQLRWKHHPLDNKDTFYVNLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGMMTEYYHFIFTTLDLYALDLEPYRYSGVNLTGFRILNVDNPHVSAIVEKWSMERLQAAPRSESGLLDGVMMTDAALLYDAVHIVSVCYQRAPQMTVNSLQCHRHKAWRFGGRFMNFIKEAQWEGLTGRIVFNKTSGLRTDFDLDIISLKEDGLEKVGVWSPADGLNITEVAKGRGPNVTDSLTNRSLIVTTVLEEPFVMFRKSDRTLYGNDRFEGYCIDLLKELAHILGFSYEIRLVEDGKYGAQDDKGQWNGMVKELIDHKADLAVAPLTITHVREKAIDFSKPFMTLGVSILYRKPNGTNPSVFSFLNPLSPDIWMYVLLAYLGVSCVLFVIARFSPYEWYDAHPCNPGSEVVENNFTLLNSFWFGMGSLMQQGSELMPKALSTRIIGGIWWFFTLIIISSYTANLAAFLTVERMESPIDSADDLAKQTKIEYGAVKDGATMTFFKKSKISTFEKMWAFMSSKPSALVKNNEEGIQRALTADYALLMESTTIEYVTQRNCNLTQIGGLIDSKGYGIGTPMGSPYRDKITIAILQLQEEDKLHIMKEKWWRGSGCPEEENKEASALGIQKIGGIFIVLAAGLVLSVLVAVGEFVYKLRKTAEREQRSFCSTVADEIRFSLTCQRRVKHKPQPPMMVKTDAVINMHTFNDRRLPGKDSMACSTSLAPVFPSpecifications
| GRIK3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_000831 | |
| GRIK3 | |
| GRIK3 (NP_000822.2, 1 a.a. ∼ 919 a.a) full-length human protein. | |
| RUO | |
| 2899 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| glutamate receptor, ionotropic, kainate 3 | |
| EAA5/GLR7/GLUR7/GluR7a | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |