Learn More
BTBD3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00022903-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MVDDKEKNMKCLTFFLMLPETVKNRSKKSSKKANTSSSSSNSSKLPPVCYEIITLKTKKKKMAADIFPRKKPANSSSTSVQQYHQQNLSNNNLIPAPNWQGLYPTIRERNAMMFNNDLMADVHFVVGPPGGTQRLPGHKYVLAVGSSVFHAMFYGELAEDKDEIRIPDVEPAAFLAMLKYIYCDEIDLAADTVLATLYAAKKYIVPHLARACVNFLETSLSAKNACVLLSQSCLFEEPDLTQRCWEVIDAQAELALKSEGFCDIDFQTLESILRRETLNAKEIVVFEAALNWAEVECQRQDLALSIENKRKVLGKALYLIRIPTMALDDFANGAAQSGVLTLNETNDIFLWYTAAKKPELQFVSKARKGLVPQRCHRFQSCAYRSNQWRYRGRCDSIQFAVDKRVFIAGFGLYGSSCGSAEYSAKIELKRQGVVLGQNLSKYFSDGSSNTFPVWFEYPVQIEPDTFYTASVILDGNELSYFGQEGMTEVQCGKVTVQFQCSSDSTNGTGVQGGQIPELIFYASpecifications
| BTBD3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_014962 | |
| BTBD3 | |
| BTBD3 (NP_055777.1, 1 a.a. ∼ 522 a.a) full-length human protein. | |
| RUO | |
| 22903 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| BTB (POZ) domain containing 3 | |
| KIAA0952/MGC130038/MGC130039/dJ742J24.1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |