Learn More
STX2, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00002054-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MRDRLPDLTACRKNDDGDTVVVVEKDHFMDDFFHQVEEIRNSIDKITQYVEEVKKNHSIILSAPNPEGKIKEELEDLNKEIKKTANKIRAKLKAIEQSFDQDESGNRTSVDLRIRRTQHSVLSRKFVEAMAEYNEAQTLFRERSKGRIQRQLEITGRTTTDDELEEMLESGKPSIFTSDIISDSQITRQALNEIESRHKDIMKLETSIRELHEMFMDMAMFVETQGEMINNIERNVMNATDYVEHAKEETKKAIKYQSKARRKLMFIIICVIVLLVILGIILATTLSSpecifications
| STX2 | |
| Polyclonal | |
| No additive | |
| NM_001980.2 | |
| STX2 | |
| STX2 (NP_001971.2, 1 a.a. ∼ 287 a.a) full-length human protein. | |
| RUO | |
| 2054 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| syntaxin 2 | |
| EPIM/EPM/MGC51014/STX2A/STX2B/STX2C | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |