Learn More
MAGED2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00010916-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MSDTSESGAGLTRFQAEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKVSHVADTKVNTKAQETEAAPSQAPADEPEPESAAAQSQENQDTRPKVKAKKARKVKHLDGEEDGSSDQSQASGTTGGRRVSKALMASMARRASRGPIAFWARRASRTRLAAWARRALLSLRSPKARRGKARRRAAKLQSSQEPEAPPPRDVALLQGRANDLVKYLLAKDQTKIPIKRSDMLKDIIKEYTDVYPEIIERAGYSLEKVFGIQLKEIDKNDHLYILLSTLEPTDAGILGTTKDSPKLGLLMVLLSIIFMNGNRSSEAVIWEVLRKLGLRPGIHHSLFGDVKKLITDEFVKQKYLDYARVPNSNPPEYEFFWGLRSYYETSKMKVLKFACKVQKKDPKEWAAQYREAMEADLKAAAEAAAEAKARAEIRARMGIGLGSENAAGPCNWDEADIGPWAKARIQAGAEAKAKAQESGSASTGASTSTNNSASASASTSGGFSAGASLTATLTFGLFAGLGGAGASTSGSSGACGFSYKSpecifications
| MAGED2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_014599.4 | |
| MAGED2 | |
| MAGED2 (NP_055414.2, 1 a.a. ∼ 606 a.a) full-length human protein. | |
| RUO | |
| 10916 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| melanoma antigen family D, 2 | |
| 11B6/BCG1/HCA10/JCL-1/MAGE-D2/MAGED/MGC8386 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |