Learn More
GPT2, Mouse anti-Human, Clone: 7A11, Abnova™
Mouse Monoclonal Antibody
Brand:  Abnova H00084706-M04.100ug
Product Code. 16176033
  • £303.00 / 100µg
Description
Sequence: NLHPEIKGQLVKLLSVRLCPPVSGQAAMDIVVNPPVAGEESFEQFSREKESVLGNLAKKAKLTEDLFNQVPGIHCNPLQGAMYAFPRIFIPAKAVEAAQ
Specifications
Specifications
GPT2
Monoclonal
Unconjugated
glutamic pyruvate transaminase (alanine aminotransferase) 2
ALT2
Mouse
100 μg
Primary
Human
Antibody
ELISA, Immunofluorescence, Western Blot
7A11
In 1x PBS, pH 7.4
NM_133443
GPT2
GPT2 (NP_597700, 358 a.a. ∼ 456 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
84706
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ