Learn More
ABCC6, Mouse anti-Human, Clone: 1E6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000368-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRVSpecifications
| ABCC6 | |
| Monoclonal | |
| Unconjugated | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 6 | |
| ABC34/ARA/EST349056/MLP1/MOATE/MRP6/PXE/PXE1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E6 | |
| In 1x PBS, pH 7.4 | |
| NM_001171 | |
| ABCC6 | |
| ABCC6 (NP_001162, 831 a.a. ∼ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 368 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |