Learn More
zinc finger protein 83 (A01), Mouse anti-Human, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ZNF83.
Brand: Abnova H00055769-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MHGRKDDAQKQPVKNQLGLNPQSHLPELQLFQAEGKIYKYDHMEKSVNSSSLVSPPQRISSTVKTHISHTYECNFVDSLFTQKEKANIGTEHYKCNERGKAFHQGLHFTIHSpecifications
| zinc finger protein 83 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ZNF83. | |
| ZNF83 | |
| FLJ11015/FLJ14876/FLJ30097/FLJ90585/HPF1/MGC33853/ZNF816B | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human, Mouse | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018300 | |
| ZNF83 | |
| ZNF83 (NP_060770, 1 a.a. ∼ 111 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 55769 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |