Learn More
Abnova™ Human ZIK1 Full-length ORF (AAI03958.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16159933
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16159933
Brand: Abnova™ H00284307P01.25ug
Used for AP, Array, ELISA, WB-Re
Sequence: MCVPVLKDILHLADLPGQKPYLVGECTNHHQHQKHHSAKKSLKRDMDRASYVKCCLFCMSLKPFRKWEVGKDLPAMLRLLRSLVFPGGKKPGTITECGEDIRSQKSHYKSGECGKASRHKHTPVYHPRVYTGKKLYECSKCGKAFRGKYSLVQHQRVHTGERPWECNECGKFFSQTSHLNDHRRIHTGERPYECSECGKLFRQNSSLVDHQKIHTGARPYECSQCGKSFSQKATLVKHQRVHTGERPYKCGECGNSFSQSAILNQHRRIHTGAKPYECGQCGKSFSQKATLIKHQRVHTGERPYKCGDCGKSFSQSSILIQHRRIHTGARPYECGQCGKSFSQKSGLIQHQVVHTGERPYECNKCGNSFSQCSSLIHHQKCHNT
Specifications
Accession Number AAI03958.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 284307
Molecular Weight (g/mol) 70.1kDa
Name ZIK1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MCVPVLKDILHLADLPGQKPYLVGECTNHHQHQKHHSAKKSLKRDMDRASYVKCCLFCMSLKPFRKWEVGKDLPAMLRLLRSLVFPGGKKPGTITECGEDIRSQKSHYKSGECGKASRHKHTPVYHPRVYTGKKLYECSKCGKAFRGKYSLVQHQRVHTGERPWECNECGKFFSQTSHLNDHRRIHTGERPYECSECGKLFRQNSSLVDHQKIHTGARPYECSQCGKSFSQKATLVKHQRVHTGERPYKCGECGNSFSQSAILNQHRRIHTGAKPYECGQCGKSFSQKATLIKHQRVHTGERPYKCGDCGKSFSQSSILIQHRRIHTGARPYECGQCGKSFSQKSGLIQHQVVHTGERPYECNKCGNSFSQCSSLIHHQKCHNT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC119699/MGC119700/MGC119701/ZNF762
Common Name ZIK1
Gene Symbol ZIK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid