Learn More
DIS3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00022894-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MLKSKTFLKKTRAGGVMKIVREHYLRDDIGCGAPGCAACGGAHEGPALEPQPQDPASSVCPQPHYLLPDTNVLLHQIDVLEDPAIRNVIVLQTVLQEVRNRSAPVYKRIRDVTNNQEKHFYTFTNEHHRETYVEQEQGENANDRNDRAIRVAAKWYNEHLKKMSADNQLQVIFITNDRRNKEKAIEEGIPAFTCEEYVKSLTANPELIDRLACLSEEGNEIESGKIIFSEHLPLSKLQQGIKSGTYLQGTFRASRENYLEATVWIHGDNEENKEIILQGLKHLNRAVHEDIVAVELLPKSQWVAPSSVVLHDEGQNEEDVEKEEERERMLKTAVSEKMLKPTGRVVGIIKRNWRPYCGMLSKSDIKESRRHLFTPADKRIPRIRIETRQASTLEGRRIIVAIDGWPRNSRYPNGHFVRNLGDVGEKETETEVLLLEHDVPHQPFSQAVLSFLPKMPWSITEKDMKNREDLRHLCICSVDPPGCTDIDDALHCRELENGNLEVGVHIADVSHFIRPGNALDQESARRGTTVYLCEKRIDMVPELLSSNLCSLKCDVDRLAFSCIWEMNHNAEILKTKFTKSVINSKASLTYAEAQLRIDSANMNDDITTSLRGLNKLAKILKKRRIEKGALTLSSPEVRFHMDSETHDPIDLQTKELRETNSMVEEFMLLANISVAKKIHEEFSEHALLRKHPAPPPSNYEILVKAARSRNLEIKTDTAKSLAESLDQAESPTFPYLNTLLRILATRCMMQAVYFCSGMDNDFHHYGLASPIYTHFTSPIRRYADVIVHRLLAVAIGADCTYPELTDKHKLADICKNLNFRHKMAQYAQRASVAFHTQLFFKSKGIVSEEAYILFVRKNAIVVLIPKYGLEGTVFFEEKDKPNPQLIYDDEIPSLKIEDTVFHVFDKVKVKIMLDSSNLQHQKIRMSLVEPQIPGISIPTDTSNMDLNGPKKKKMKLGKSpecifications
| DIS3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC056143.1 | |
| DIS3 | |
| KIAA1008 (AAH56143.1, 1 a.a. ∼ 958 a.a) full-length human protein. | |
| RUO | |
| 22894 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| DIS3 mitotic control homolog (S. cerevisiae) | |
| DKFZp667L1817/EXOSC11/FLJ10484/KIAA1008/MGC33035/RP11-342J4.3/RRP44/bA555G22.1/dis3p | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |