Learn More
UBTD2, Mouse anti-Human, Clone: 1B8-1B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00092181-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MTDGQLRSKRDEFWDTAPAFEGRKEIWDALKAAAHAFESNDHELAQAIIDGANITLPHGALTECYDELGNRYQLPVYCLAPPINMIEEKSDIETLDIPEPPPNSGYECQLRLRLSTGKDLKLVVRSTDTVFHMKRRLHAAEGVEPGSQRWFFSGRPLTDKMKFEELKIPKDYVVQVIVSQPVQNPTPVENSpecifications
| UBTD2 | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin domain containing 2 | |
| DC-UbP/DCUBP/MGC30022 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1B8-1B1 | |
| In 1x PBS, pH 7.4 | |
| BC019910 | |
| UBTD2 | |
| DC-UbP (AAH19910, 1 a.a. ∼ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 92181 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |