Learn More
Abnova™ Human KLF1 Partial ORF (NP_006554, 183 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16155532
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16155532
Brand: Abnova™ H00010661Q01.10ug
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for AP, Array, ELISA, WB-Re
Sequence: PRTGLSVPAASGAPYGLLSGYPAMYPAPQYQGHFQLFRGLQGPAPGPATSPSFLS
Specifications
Accession Number NP_006554
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10661
Molecular Weight (g/mol) 31.79kDa
Name KLF1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen PRTGLSVPAASGAPYGLLSGYPAMYPAPQYQGHFQLFRGLQGPAPGPATSPSFLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EKLF
Common Name KLF1
Gene Symbol KLF1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid