Learn More
Abnova™ Human PGM5 Full-length ORF (ENSP00000366540, 1 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16141268
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
Product Code. 16141268
Brand: Abnova™ H00005239P01.10ug
Used for AP, Array, ELISA, WB-Re
Phosphoglucomutases (EC 5.2.2.2.), such as PGM5, are phosphotransferases involved in interconversion of glucose-1-phosphate and glucose-6-phosphate. PGM activity is essential in formation of carbohydrates from glucose-6-phosphate and in formation of glucose-6-phosphate from galactose and glycogen (Edwards et al., 1995 [PubMed 8586438]).[supplied by OMIM]
Sequence: MVVGSDGRYFSRTAIEIVVQMAAANGIGRLIIGQNGILSTPAVSCIIRKIKAAGGIILTASHCPGGPGGEFGVKFNVANGGPAPDVVSDKIYQISKTIEEYAICPDLRIDLSRLGRQEFDLENKFKPFRVEIVDPVDIYLNLLRTIFDFHAIKGLLTGPSQLKIRIDAMHGVMGPYVRKVLCDELGAPANSAINCVPLEDFGGQHPDPNLTYATTLLEAMKGGEYGFGAAFDADGDRYMILGQNGFFVSPSDSLAIIAANLSCIPYFRQMGVRGFGRSMPTSMALDRVAKSMKVPVYETPAGWRFFSNLMDSGRCNLCGEESFGTGSDHLREKDGLWAVLVWLSIIAARKQSVEEIVRDHWAKFGRHYYCRFDYEGLDPKTTYYIMRDLEALVTDKSFIGQQFAVGSHVYSVAKTDSFEYVDPVDGTVTKKQGLRIIFSDASRLIFRLSSSSGVRATLRLYAESYERDPSGHDQEPQAVLSPLIAIALKISQIHERTGRRGPTVIT
Specifications
Accession Number ENSP00000366540
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5239
Molecular Weight (g/mol) 82kDa
Name PGM5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MVVGSDGRYFSRTAIEIVVQMAAANGIGRLIIGQNGILSTPAVSCIIRKIKAAGGIILTASHCPGGPGGEFGVKFNVANGGPAPDVVSDKIYQISKTIEEYAICPDLRIDLSRLGRQEFDLENKFKPFRVEIVDPVDIYLNLLRTIFDFHAIKGLLTGPSQLKIRIDAMHGVMGPYVRKVLCDELGAPANSAINCVPLEDFGGQHPDPNLTYATTLLEAMKGGEYGFGAAFDADGDRYMILGQNGFFVSPSDSLAIIAANLSCIPYFRQMGVRGFGRSMPTSMALDRVAKSMKVPVYETPAGWRFFSNLMDSGRCNLCGEESFGTGSDHLREKDGLWAVLVWLSIIAARKQSVEEIVRDHWAKFGRHYYCRFDYEGLDPKTTYYIMRDLEALVTDKSFIGQQFAVGSHVYSVAKTDSFEYVDPVDGTVTKKQGLRIIFSDASRLIFRLSSSSGVRATLRLYAESYERDPSGHDQEPQAVLSPLIAIALKISQIHERTGRRGPTVIT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PGMRP
Common Name PGM5
Gene Symbol PGM5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid