Learn More
OTC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OTC.
Brand: Abnova PAB19978.100uL
Description
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also. [provided by RefSeq
Sequence: QLKGRDLLTLKNFTGEEIKYMLWLSADLKFRIKQKGEYLPLLQGKSLGMIFEKRSTRTRLSTETGFALLGGHPCFLTTQDIHLGVNESLTDTARVLSSMADAVLARSpecifications
| OTC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OTC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC129967/MGC129968/MGC138856/OCTD | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5009 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| OTC | |
| OTC | |
| Recombinant protein corresponding to amino acids of human OTC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |