Learn More
GADD45G, Mouse anti-Human, Clone: 1D3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010912-M01.50ug
This item is not returnable.
View return policy
Description
Sequence: MTLEEVRGQDTVPESTARMQGAGKALHELLLSAQRQGCLTAGVYESAKVLNVDPDNVTFCVLAAGEEDEGDIALQIHFTLIQAFCCENDIDIVRVGDVQRLAAIVGAGEEAGAPGDLHCILISNPNEDAWKDPALEKLSLFCEESRSVNDWVPSITLPESpecifications
| GADD45G | |
| Monoclonal | |
| Unconjugated | |
| growth arrest and DNA-damage-inducible, gamma | |
| CR6/DDIT2/GADD45gamma/GRP17 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1D3 | |
| In 1x PBS, pH 7.4 | |
| BC019325 | |
| GADD45G | |
| GADD45G (AAH19325, 1 a.a. ∼ 159 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10912 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |