Learn More
WBP2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand:  Abnova H00023558-B02P.50ug
Product Code. 16114826
  • £303.00 / 50µg
Description
Sequence: MALNKNHSEGGGVIVNNTESILMSYDHVELTFNDMKNVPEAFKGTKKGTVYLTPYRVIFLSKGKDAMQSFMMPFYLMKDCEIKQPVFGANYIKGTVKAEAGGGWEGSASYKLTFTAGGAIEFGQRMLQVASQASRGEVPSGAYGYSYMPSGAYVYPPPVANGMYPCPPGYPYPPPPPEFYPGPPMMDGAMGYVQPPPPPYPGPMEPPVSGPDVPSTPAAEAKAAEAAASAYYNPGNPHNVYMPTSQPPPPPYYPPEDKKTQ
Specifications
Specifications
WBP2
Polyclonal
In 1x PBS, pH 7.4
NM_012478.3
WBP2
WBP2 (NP_036610.2, 1 a.a. ∼ 261 a.a) full-length human protein.
RUO
23558
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Western Blot
Unconjugated
WW domain binding protein 2
MGC18269/WBP-2