Learn More
ZBTB45, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZBTB45.
Brand:  Abnova PAB22695.100uL
Product Code. 16105370
  • £535.00 / 100µL
Description
Sequence: PTLQPEAAPSTQLGEVPAPSAAPTTAPSGTPARTPGAEPPTYECSHCRKTFSSRKNYTKHMFIHSGEKPHQCAVCW
Specifications
Specifications
ZBTB45
Polyclonal
Rabbit polyclonal antibody raised against recombinant ZBTB45.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
DKFZp547H249/FLJ14486/ZNF499
Rabbit
Antigen affinity purification
RUO
84878
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
ZBTB45
ZBTB45
Recombinant protein corresponding to amino acids of human ZBTB45.