Learn More
SORCS3, Mouse anti-Human, Clone: 3B12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00022986-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: KRKIPWINIYAQVQHDKEQEMIGSVSQSENAPKITLSDFTEPEELLDKELDTRVIGGIATIANSESTKEIPNCTSVSpecifications
| SORCS3 | |
| Monoclonal | |
| Unconjugated | |
| sortilin-related VPS10 domain containing receptor 3 | |
| SORCS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B12 | |
| In 1x PBS, pH 7.4 | |
| NM_014978 | |
| SORCS3 | |
| SORCS3 (NP_055793, 1147 a.a. ∼ 1222 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 22986 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |