Learn More
TBX2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand:  Abnova H00006909-B02P.50ug
Product Code. 16103438
  • £303.00 / 50µg
Description
Sequence: MAYHPFHAPRPADFPMSAFLAAAQPSFFPALALPPGALAKPLPDPGLAGAAAAAAAAAAAAEAGLHVSALGPHPPAAHLRSLKSLEPEDEVEDDPKVTLEAKELWDQFHKLGTEMVITKSGRRMFPPFKVRVSGLDKKAKYILLMDIVAADDCRYKFHNSRWMVAGKADPEMPKRMYIHPDSPATGEQWMAKPVAFHKLKLTNNISDKHGFTILNSMHKYQPRFHIVRANDILKLPYSTFRTYVFPETDFIAVTAYQNDKITQLKIDNNPFAKGFRDTGNGRREKRKQLTLPSLRLYEEHCKPERDGAESDASSCDPPPAREPPTSPGAAPSPLRLHRARAEEKSCAADSDPEPERLSEERAGAPLGRSPAPDSASPTRLTEPERARERRSPERGKEPAESGGDGPFGLRSLEKERAEARRKDEGRKEAAEGKEQGLAPLVVQTDSASPLGAGHLPGLAFSSHLHGQQFFGPLGAGQPLFLHPGQFTMGPGAFSAMGMGHLLASVAGGGNGGGGGPGTAAGLDAGGLGPAASAASTAAPFPFHLSQHMLASQGIPMPTFGGLFPYPYTYMAAAAAAASALPATSAAAAAAAAAGSLSRSPFLGSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGLESQRALSPGRESPK
Specifications
Specifications
TBX2
Polyclonal
In 1x PBS, pH 7.4
NM_005994.3
TBX2
TBX2 (NP_005985.2, 1 a.a. ∼ 702 a.a) full-length human protein.
RUO
6909
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Western Blot
Unconjugated
T-box 2
FLJ10169