Learn More
PLEKHO1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PLEKHO1.
Brand: Abnova PAB16843.100ug
This item is not returnable.
View return policy
Specifications
| PLEKHO1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PLEKHO1. | |
| In PBS (0.02% sodium azide) | |
| CKIP-1/OC120 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PLEKHO1 | |
| PLEKHO1 | |
| A synthetic peptide corresponding to C-terminus 18 amino acids of human PLEKHO1. | |
| RUO | |
| 51177 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |