Learn More
ABCC10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ABCC10.
Brand: Abnova PAB25110.100uL
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi-drug resistance. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq
Specifications
| ABCC10 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| ABCC10 | |
| ABCC10 | |
| A synthetic peptide corresponding to ABCC10. | |
| 100 μL | |
| Primary | |
| ABCC10 polyclonal antibody detects endogenous levels of ABCC10 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ABCC10. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| EST182763/MRP7/SIMRP7 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 89845 | |
| Human | |
| Liquid |