Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-3 Protein
Product Code. 15999578
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Product Code. 15999578
Brand: enQuireBio™ QP12169100ug
A cDNA sequence encoding the HCV NS3 Genotype-3 was constructed and used to recombinantly synthesize the protein.
Specifications
Name Hepatitis C HCV NS3 Genotype-3 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Hepatitis C
Species E. coli
Protein Tag GST
Buffer 1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
Purity or Quality Grade HCV NS3 Genotype-3 protein is >95% pure as determined by 10% PAGE (coomassie staining).