Learn More
Invitrogen™ MAG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579620
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. IHC: mouse brain tissue, rat cerebellum tissue.
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
| MAG | |
| Polyclonal | |
| Unconjugated | |
| MAG | |
| 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17136, 29409, 4099 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P07722, P20916, P20917 | |
| MAG | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human MAG (114-132aa KYYFRGDLGGYNQYTFSEH). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |