Learn More
Invitrogen™ NEU2 Monoclonal Antibody (OTI4F4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525555
Product Code. 15975154
  • £475.00 / 100µL
Description
NEU2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NEU2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y3R4
NEU2
Full length human recombinant protein of NEU2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Western Blot, Immunocytochemistry
OTI4F4
Unconjugated
NEU2
Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); SIAL2; sialidase 2 (cytosolic sialidase); sialidase-2
Mouse
Affinity Chromatography
RUO
4759
-20°C, Avoid Freeze/Thaw Cycles