Learn More
Invitrogen™ PKNOX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565946
This item is not returnable.
View return policy
Description
Immunogen sequence: DASNPDPAPKA KKIKSQHRPT QRFWPNSIAA GVLQQQGGAP GTNPDGSINL DNLQSLS Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
PREP-2 (Pbx-regulating protein-2), also known as PBX/knotted 1 homeobox 2 or PKNOX2, is a widely expressed protein belonging to the TALE (three amino acid loop extension)/MEIS family. PREP-2 is a DNA-binding protein that forms stable complexes with Pbx proteins. It is highly homologous to the related protein PREP-1, but displays a more restricted tissue distribution and a higher DNA-dissociation rate. Like PREP-1, PREP-2 forms a heterodimer with Pbx 1. The PREP-2-Pbx 1 dimer is relocated to the nucleus where it associates with HoxB1 to form a ternary complex. In contrast with PREP-1, which acts to increase transcriptional activation in this ternary complex, PREP-2 leads to a slight decrease in transcriptional activity of the ternary complex. Multiple isoforms exist for PREP-2, localizing to the nucleus or cytoplasm. Cytoplasmic isoforms are believed to colocalize with F-actin, G-actin and tubulin/microtubules.
Specifications
| PKNOX2 | |
| Polyclonal | |
| Unconjugated | |
| PKNOX2 | |
| D230005H23Rik; homeobox protein Meis2-like; homeobox protein PKNOX2; Homeobox protein PREP-2; homeodomain containing transcription factor PREP2; Pbx/knotted 1 homeobox 2; PBX/knotted homeobox 2; Pknox2; PREP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63876 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96KN3 | |
| PKNOX2 | |
| Recombinant Human PKNOX2. Recombinant protein control fragment (Product #RP-106627). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |